Learn More
Carolina™ Individual Bacterial Cultures (D-M)
Bacteria cultures have wide applications in the food and chemical industries because of their highly diverse metabolic activities.
Specifications
For Use With (Application) | Ideal for food and chemical industries because of their highly diverse metabolic activities. |
---|---|
Storage Requirements | 20°C to 22°C |
Quantity | Ea. |
Includes | Carolina™ techniques for studying bacteria and fungi manual |
Sufficient For | Class of 30 students |
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog Number | Mfr. No. | Product Type | Media Type | Scientific Name | Temperature (Metric) Range | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Media Type | Scientific Name | Temperature (Metric) Range | Price | Quantity & Availability | ||
S07417ND
|
Carolina Biological Supply
155068 |
Bacteria Culture | Nutrient Agar | E.coli | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07427ND
|
Carolina Biological Supply
155155A |
Bacteria MicroKwik Culture™ | Brain Heart Infusion Agar/Broth | Micrococcus luteus | 25°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07210ND
|
Carolina Biological Supply
155160 |
Bacteria Culture | Nutrient Agar | Micrococcus roseus | 25°C |
Each for $15.00
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S07408ND
|
Carolina Biological Supply
155600A |
Bacteria MicroKwik Culture™ | Dextrose Starch Agar or Brain Heart Infusion Agar | Enterococcus faecalis | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07418ND
|
Carolina Biological Supply
155067 |
Bacteria Culture | Nutrient Agar | E.coli | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07192ND
|
Carolina Biological Supply
155155 |
Bacteria Culture | Brain Heart Infusion Agar/Broth | Micrococcus luteus | 25°C |
Each for $15.00
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S07413ND
|
Carolina Biological Supply
155095A |
Bacteria MicroKwik Culture™ | Nutrient Agar | Klebsiella pneumoniae | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07180ND
|
Carolina Biological Supply
155030 |
Bacteria Culture | Nutrient Agar | Enterobacter aerogenes | 30°C |
Each for $15.00
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S07193ND
|
Carolina Biological Supply
155180A |
Bacteria MicroKwik Culture™ | Brain Heart Infusion Agar | Mycobacterium smegmatis | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07407ND
|
Carolina Biological Supply
155030A |
Bacteria MicroKwik Culture™ | Nutrient Agar | Enterobacter aerogenes | 30°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07404ND
|
Carolina Biological Supply
155610A |
Bacteria MicroKwik Culture™ | Tomato Juice, Yeast Extract, Milk Broth or Brain Heart Infusion Agar | Lactococcus lactus | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07194ND
|
Carolina Biological Supply
155170A |
Bacteria MicroKwik Culture™ | Brain Heart Infusion Agar | Mycobacterium phlei | 37°C |
Each for $23.25
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S07403ND
|
Carolina Biological Supply
155065A |
Bacteria MicroKwik Culture™ | Nutrient Agar | E.coli | 37°C |
Each for $23.25
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S07189ND
|
Carolina Biological Supply
155065 |
Bacteria Culture | Nutrient Agar | E.coli | 37°C |
Each for $15.00
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Bacteria are involved in many of the events that make life possible for other organisms, including humans. Fossils of prokaryotes similar to today's bacteria have been found in 3.5 billion-year-old rocks. A mastery of bacteria is the key to understanding life on earth in the past, present, and future.
- Labeled by genus and species name and media on which they are cultured
- All Gram staining based on 24 hr. cultures
- Pure and high-quality
- For every level of study
- Easy-to-use media
Specifications
Ideal for food and chemical industries because of their highly diverse metabolic activities. | |
Ea. | |
Class of 30 students |
20°C to 22°C | |
Carolina™ techniques for studying bacteria and fungi manual |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.