Learn More
Carolina™ Formula 4-24™ Drosophila Medium
This medium allows teachers to set up a new culture in less than a minute.
$31.50 - $136.00
Specifications
For Use With (Application) | Medium allows teachers to set up a new culture in less than a minute. |
---|---|
Includes | Yeast, measuring cups and instructions |
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Includes:
- Yeast
- Measuring cups
- Instructions
Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity & Availability | ||||
S07691ND
|
Carolina™
173212 |
Blue Formula 4-24™ Instant Medium | 4L |
Each for $33.75
|
|
||||
S07690ND
|
Carolina™
173204 |
Plain Formula 4-24™ Instant Medium | 4 x 4L |
Case of 4 for $136.00
|
|
||||
S07689ND
|
Carolina™
173202 |
Plain Formula 4-24™ Instant Medium | 4L |
Each for $31.50
|
|
||||
S07692ND
|
Carolina™
173214 |
Blue Formula 4-24™ Instant Medium | 4 x 4L |
Case of 4 for $115.00
|
|
||||
Description
- Mold resistant
- Needs no cooking or sterilizing
- Just add water for a complete medium
- Each liter of Formula 4-24™ will prepare 75 to 85
- Drosophila culture vials
Specifications
Medium allows teachers to set up a new culture in less than a minute. | |
Yeast, measuring cups and instructions |