Learn More
Carolina™ Bran Meal
Use to culture mealworms and darkling beetles.
Supplier: Carolina™ 144322
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Description
- 5 kg package
- Great for culturing mealworms
Specifications
Food for Tenebrio molitor | |
Used for culturing darkling beetles and mealworms. | |
5 kg | |
Non-sterile |