Learn More
Sklar Instruments Cardiology Stethoscope, Sklar 27 in., Single Head, Single Head. It measures 27 inches overall and has black PVC tubing.
Supplier: Sklar Instruments 062750
Cardiology Stethoscope, Sklar. 27 in., Single Head, Single Head. Sklar's Low Profile Cardiology Stethoscope features a single, zinc alloy chestpiece. The low profile design allows the user to apply greater pressure to the diaphragm, creating a better acoustic seal resulting in more accurate identification of what is being heard. The cardiology stethoscope also has thicker, shorter 2-in-1 tubing compared to a standard stethoscope. This provides a shorter distance for the sounds to travel and decreases interference of curled tubing that can occur with longer stethoscopes. Sklar's Low Profile Cardiology Stethoscope allows the user to hear both high and low frequency sounds, specifically those identified with the heart such as blood flow, clicks, and murmurs. This stethoscope comes with 2 additional sets of ear tips and 1 additional diaphragm. It measures 27 inches overall and has black PVC tubing.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.