Learn More
Cardinal Health™ Sterile Micro-Clean ™ Sleeves
Click to view available options
Micro-Clean Sleeve
Description
Disposable sleeves have a Sterility Assurance Level of 10-6.
Lot number is embossed on each package and garment.
- Made of sterile Tyvek
- Feature bound seams and tunnelized elastic to eliminate raw edges
- Length: 18 in. (45.7cm)
- Color: white
Order Info
Packaged under ISO Class 5 (formerly Class 100) conditions. Pair packaged.
Specifications
Material | Tyvek |
Model | A6128-26 |