Learn More
Electron Microscopy Sciences Aurion Carboxyl Functionalized Gold Nanoparticles, 10 nm 5 ML
Supplier: Electron Microscopy Sciences 25601
This product has been developed to facilitate conjugating gold nanoparticles to molecules that cannot be conjugated via the classic direct adsorption method. The product is especially suited for covalent conjugation of small ligands. Aurion Gold Nanoparticles (Carboxyl Functionalized) are polyethylene glycol-carboxyl stabilized gold nanoparticle solutions based on defined particle sizes of 6, 10, 15 and 25nm.
Particle size: 10 nm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.