Learn More
Thermo Scientific CARBOPAC PA1 GUARD COLOMN
Supplier: Thermo Scientific 043096
Standard bore gaurd. For isocratic determinations of monosaccharides, disaccharides, and linear polysaccharides in a variety of matrices, use the rugged Thermo Scientifica, c Dionexa, c CarboPaca, c Pa1 analytical & Guard Columns. This specialized anion-exchange column, combined with pulsed amperometric detection (PaD), delivers high-resolution separations. The unique Thermo Scientifica, c Dionexa, c MicroBeada, c pellicular structure of the Dionex CarboPac Pa1 resin gives it stability from pH 0-14 at all concentrations of buffer salts. The pellicular resin structure provides excellent mass transfer resulting in fast gradient re-equilibration.