Learn More
OHAUS Carbon Steel Defender 3000 Series Floor Scale
Defender 3000 Floor Platforms meet dry weighing needs for industrial and commercial use. Their high-quality construction is designed to minimize flexing under heavy loads for years of dependable performance. The indicator features a bright display that changes color when in check mode. Scale also includes counting and totalization modes.
$1934.00 - $2299.00
Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
01-920-185
|
OHAUS
30837693 |
i-DF33XW5000B1L |
Each for $2,299.00
|
|
|||||
01-920-186
|
OHAUS
30837692 |
i-DF33XW2500B1L |
Each for $2,299.00
|
|
|||||
01-920-187
|
OHAUS
30837691 |
i-DF33XW2500B1R |
Each for $2,038.00
|
|
|||||
01-920-191
|
OHAUS
30837604 |
i-DF33P5000B1L |
Each for $2,195.00
|
|
|||||
01-920-192
|
OHAUS
30837603 |
i-DF33P2500B1L |
Each for $2,195.00
|
|
|||||
01-920-194
|
OHAUS
30837601 |
i-DF33P2500B1R |
Each for $1,934.00
|
|
|||||
Description
- Painted carbon steel treaded decks with four alloy steel load cells, fixed top plate (5 mm/0.196 in. thick) with structural bracing for stiff construction.
- Platforms available in sizes of 3 x 3 ft. , 4 x 4 ft. , and 5 x 5 ft. and capacities from 2,500 lb. to 5,000 lb
- Results are clearly shown on the large, bright backlit LCD display.
- In checkweighing mode, display changes color (yellow/green/red) to alert users when within the target weight range.
Specifications
Scale | |
Class III |