Learn More
Ted Pella Inc Carbon Film only on 200 mesh, Copper, 25/pk
Supplier: Ted Pella Inc 01840
Carbon films with a thickness of 15-25nm with no Formvar used during manufacturing. Completely free of Formvar. Carbon films are thin and highly transparent to electrons. They exhibit very fine grain and minimal interference with specimen structure. Films are ready to use upon receipt. The pure carbon films tend to be more delicate than support films with a Formvar backing and require more delicate handling during specimen preparation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.