Learn More
Fuel Cell Earth Carbon Cloth Plain 40cm x 40cm
Supplier: Fuel Cell Earth CCP40
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Carbon ClothGraphitized Spun Yarn Carbon FabricsWOVEN CARBON FABRICS are carbon fiber fabrics made from spun yarn. Fabric tailorability results from controlling the yield on rovings and yarns, and allows for a variety of finished composite thicknesses.The spun yarns have many surface fibrils that protrude in various directions at various lengths from thesurface. This fiber loft contributes to high cross ply tensiles and interlaminar shear strengths for 2 Dcomposite materials. Contour conformation without wrinkling is another advantage of spun yarn fabrics.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.