Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Safety Apparel™ Carbon Armour™ AV Base Layer Pants
Click to view available options
Size:
2X-Large
3X-Large
Large
Medium
Small
X-Large
Description
- CAT 2
- Fabric/Material: 6 oz. Carbon Armour AV
- FR fabric will not melt, drip, or ignite
- Unmatched comfort and protection
- Moisture wicking and quick drying
- Rib knit cuffs
- Made in the USA
Specifications
Size | 3X-Large |
Color | Gray |
Arc Flash Rating | 12 cal/cm2 |
Certifications/Compliance | NFPA 70E for Arc Flash, ASTM F1506 for Arc Flash, CSA Z462 for Arc Flash (Canada), ASTM F1959, OSHA 1910.269 |
For Use With (Application) | FR base layer |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction