Learn More
National Safety Apparel™ Carbon Armour™ AV Base Layer Shirt
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- CAT 2
- Fabric/Material: 6 oz. Carbon Armour AV
- FR fabric will not melt, drip, or ignite
- Mock turtleneck collar
- Unmatched comfort and protection
- Moisture wicking and fast drying
- Rib knit cuffs
- Made in the USA
Specifications
Size | Small |
Color | Gray |
Arc Flash Rating | 12 cal/cm2 |
Certifications/Compliance | NFPA 70E for Arc Flash, ASTM F1506 for Arc Flash, CSA Z462 for Arc Flash (Canada), ASTM F1959, OSHA 1910.269 |