Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon, Activated (Granular), Fisher Chemical

Catalog Number c270c
Click to view available options
Quantity:
350 g
Packaging:
Glass Bottle
7440-44-0
C
12.011
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
5462310
CHEBI:27594
carbon
[C]
7440-44-0
C
12.011
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
5462310
CHEBI:27594
carbon
[C]

Grayish-black granules

TRUSTED_SUSTAINABILITY
Melting Point 3652°C
Color Black
Density 1.8g/mL
Packaging Glass Bottle
Quantity 350 g
Vapor Pressure 1mmHg at 3586°C
Physical Form Solid
Chemical Name or Material Carbon, Activated
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin and eye irritation. Avoid contact with skin and eyes. Use personal protective equipment. Look out! DATATYPE 710. Avoid breathing dust/fume/gas/mist/vapours/spray. Rinse with water. . Obtain medical attention. Remove from exposure, lie down. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. If not breathing, give artificial respiration. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:0
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit