Learn More
Pfanstiehl Inc a. a-Trehalose Dihydrate. High Purity (Low Endotoxin) NF. EP. JP. ChP (T-104-4) 5kg pack GMP, Compendial CAS: 6138-23-4
Supplier: Pfanstiehl Inc T10445KG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Trehalose is a non-reducing disaccharide consisting of two glucose molecules linked by an , 1,1 bond. Pfanstiehl, High Purity Low Endotoxin Trehalose is used in the biopharmaceutical industry to stabilize proteins, lipids, and carbohydrates throughout the formulation and freeze/thaw life cycle of therapeutics. It is also widely applied as a key component in cryopreservation media to stabilize cell membranes and improve the recovery and robustness of cells used for bioproduction and cell therapy.The stabilizing benefits of trehalose include but are not limited to: Monoclonal Antibodies (mAbs) Antibody Fragments (fAbs) Vaccines Fusion Proteins Peptides Stem cellsPfanstiehl Brand Trehalose is available as NF (National Formulary), EP (European Pharmacopoeia), JP (Japanese Pharmacopoeia), and ChP (Chinese Pharmacopoeia).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.