Learn More
Pfanstiehl Inc D-Galactose, Lactose Derived, High Purity Low Endotoxin, NF, EP (G-125-1) – 1kg pack – GMP, Multicompendial – CAS: 59-23-4
Supplier: Pfanstiehl Inc G12511KG
Galactose is a monosaccharide sugar of the hexose class that is a constituent of lactose and many polysaccharides. Galactose is found in milk, sugar beets, gums, and mucilages. It is also synthesized in humans and is a constituent of glycolipids and glycoproteins. Galactose can be used as a cell metabolism modulator in the optimization of protein therapeutics bioproduction.
Pfanstiehl High Purity Low Endotoxin Galactose was developed specifically for biopharmaceutical use, to the highest quality and purity standards. It has been utilized as a key, chemically defined component to optimize protein production while mitigating lactate and ammonia formation. The various benefits imparted by Galactose in bioproduction include but are not limited to: Reduced Lactate & Ammonia Production; Increased Target Protein Production; Clean and Consistent Media Impurity Profile.
Pfanstiehl Galactose is available as NF (National Formulary) and EP (European Pharmacopoeia).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.