Learn More
Pfanstiehl Inc Sucrose, NF, EP, JP, ChP High Purity Low Endotoxin - Cane Derived (S-124-1-MC) – 1kg pack – GMP, Compendial – CAS: 57-50-1
Supplier: Pfanstiehl Inc S1241MC1KG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Sucrose is a non-reducing crystalline disaccharide made up of glucose and fructose, found in many plants but extracted as ordinary sugar mainly from sugar cane and sugar beets. Pfanstiehl, High Purity Low Endotoxin Sucrose is used in the biopharmaceutical industry to stabilize proteins, lipids, and carbohydrates throughout the formulation and freeze/thaw life cycle of therapeutics. It is also widely applied as a cryopreservative and media supplement in a variety of cell-based bioprocesses. It is preferred to trehalose in applications where solubility and/or viscosity issues are paramount.
The benefits of sucrose include but are not limited to: Monoclonal Antibody (mAb) Stabilization; Vaccines Stabilization; Peptide Stabilization; Antibody Drug Conjugate (ADC) Stabilization; Stem Cell Preservation; Media Supplementation
Pfanstiehl Sucrose is available as NF (National Formulary), EP (European Pharmacopoeia), JP (Japanese Pharmacopoeia), and ChP (Chinese Pharm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.