Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ CApture™ BT Fuming System Accessory, Prefilters
For use with CApture BT Fuming System
$123.00 - $258.00
Catalog Number | Mfr. No. | Filter Type | Length (Metric) | WidthMetric | Material | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Filter Type | Length (Metric) | WidthMetric | Material | Price | Quantity & Availability | ||
31-814-00
|
Labconco™
3181400 |
Cabinet Prefilter | Filter: 50.8 to 76.2 cm | Polyester |
Each for $123.00
|
|
|||
31-814-01
|
Labconco™
3181401 |
Humidifier Prefilter | Filter: 25.4 to 50.8 cm | Activated Carbon |
Each for $258.00
|
|
|||
Description
Carbon-based and HEPA filters used in variety of Labconco enclosures require periodic replacementSpot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title