Learn More
Cytiva Capto SP ImpRes Validation column
Supplier: Cytiva 29315186
Validation columns are prepacked ready-to-use Tricorn 10/200 columns suitable for scale-down studies such as viral clearance. Cation exchange technique.