Learn More
Cytiva Capto adhere Validation column
Supplier: Cytiva 29315195
Validation columns are prepacked ready-to-use Tricorn 10/200 columns suitable for scale-down studies such as viral clearance. Multimodal technique.