Learn More
Erlab™ CaptairFlow 321 Clean Air Enclosure with HEPA Filter
Laminar flow hood with HEPA filter and UV lights to eliminate contamination between operations.
Supplier: Erlab™ 321LFH
Description
Providing an ultra-clean and dust-free work area, the CaptairFlow Clean Air Enclosure is designed to prevent environmental pollution and cross-contamination. The enclosure features a high efficiency filtration system that provides a particle-free workstation around the manipulation, bringing HEPA filtered clean air down to the work surface. High efficiency UV lights are used to decontaminate the worktop from biological cross-contamination between operations. Smart Technology™ gives you an easily identifiable method of communication regarding sash status or air face velocity: check sash, pre-filter, HEPA/ULPA, and fan status via an LED band of light around the enclosure called a Smart-Light.
- For non-pathogenic cell cultures, in-vitro cultures, microbiology, homeopathic preparations, electronics, and optics
- Smart Technology™ tool consists of Smart-Light Communication, chemical sensors, real-time status, and eGuard app
- Comes equipped with needed quantity of GF4.HP filter and Pre-Filter M for first time use
- Units do not come standard with a stainless steel work surface
Specifications
Flow Hood | |
55 W | |
298.5 mm | |
99.995% | |
Protecting Samples In Clean Bench Procedures | |
CaptairFlow 321 Filter Housing, Enclosure, Fan Box, UV Light, Hardware | |
1105 to 1292 mm | |
31.6 in. | |
50/60 Hz | |
616 mm | |
68 kg | |
0.59 A |
<60 dBA | |
43.5 to 50.9 in. | |
H14 HEPA | |
GF4.HP, Pre-filter M, and Pre-filter XLS | |
Corrosion Resistant Electro-galvanized Steel Coated with Anti-acid Polymer and Polypropylene | |
LED, UV | |
11.75 in. | |
803 mm | |
24.5 in. | |
150 lb. | |
90/220 V |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.