Learn More
Electron Microscopy Sciences Capsule Mold Type A, 10 Cavities
Supplier: Electron Microscopy Sciences 70150
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Our molds are made from the strongest possible silicone rubber providing extremely high tear resistance. The molds are are reusable. Type A has 10 cavities, 8mm diameter body, 5mm diameter tip, 11mm height, tapered tip.