Learn More
Honeywell Burdick & Jackson™ Capping Reagent, 80/10/10 THF/Pyridine/Acetic Anhydride (v/v)
BioSyn™, for DNA/RNA synthesis
Supplier: Honeywell Burdick & Jackson™ BR64110
This item is not returnable.
View return policy
Description
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Specifications
109-99-9 | |
0.888 g/cm3 | |
2 x 10 L |
Room Temperature | |
HDPE NOWPAK | |
Liquid |