Learn More
Fhc Inc CAPILLARY TUBING 250/PKG
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Fhc Inc 27300
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Capillary Tubing - for Single Cell Recording and Microinjection, 250/PKG, Borosil 1.0mm OD x 0.75mm ID