Learn More
MilliporeSigma™ Supelco™ Capillary Cleaving™ Tools and Accessories
Increases column performance by making the properly sized, squarely-cut ferrule to seal capillary column ends
Supplier: MilliporeSigma™ Supelco™ 23814
Description
- Makes scalpel-like cuts to polyimide-coated fused silica
- Leaves no jagged edges to create problems
- Industrial sapphire cutting edges remains sharp indefinitely
- Spring-loaded retractable blade reduces the chance of breakage if the tool is dropped
- Replacement sapphire cutting blades are available
Specifications
Making Scalpel-Like Cuts to Polyimide-Coated Fused Silica | |
Sapphire Cutting Blade | |
Retractable Blade Version |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cleaving is a trademark of Sigma-Aldrich Co. LLC