Learn More
Waters Corp White Cap with X-Slit PTFE Silicone Septum
Supplier:  Waters Corp 186004632
Encompass_Preferred
This cap is used with the 10 ml vial for Waters 2707 Autosamper. The cap has an X-Slit PTFE Silicone Septa.
Catalog No. 50-466-016
Explore available promotions