Learn More
Waters Corp Red, 12 x 32 mm Snap Neck Cap and PTFE/Silicone Septum, 100/pk
Supplier: Waters Corp 186002650
Waters offers a selection of cap, septum, and assembled cap/septum for various vial sizes. The range of septum material is limited to PTFE, silicone / PTFE, and polyethylene septumless caps.