Learn More
Chemglass Life Sciences Cap, GC Chem Snap, Polypropylene, with PTFE/Red Rubber Septum
SDP
Supplier:  Chemglass Life Sciences CV35750011
Encompass_Preferred
Cap, GC Chem Snap, Polypropylene, with PTFE/Red Rubber Septum. GC Chem Snap Caps were developed to handle the volatile samples, providing a liquid airtight seal with the vial. Simply snap on the cap and an evaporation-proof seal of a crimped cap is achieved.
Catalog No. 50-153-2557