Learn More
Sklar Instruments Cannula Instrument Cleaning Brush, Sklar, 15mm x 18 in., Nylon / Stainless Steel. PK/3.
Supplier: Sklar Instruments 101366
Cannula Instrument Cleaning Brush, Sklar, 15mm x 18 in., Nylon / Stainless Steel. Sklar's Cannula Instrument Cleaning Brushes are used in the multiple-step process of sterilizing a reusable cannula. Following soaking in enzymatic soluntion the cannula should be cleaned manually with a brush and detergent or ultrasonically. Sklar cannula cleaning brushes are designed for manual cleaning and are available in several sizes in order to properly decontaminate specific size cannulas used for specific procedures. This product has a 15mm wide cleaning brush and an overall length of 18 inches. It also features a loop at the end of the handle for improved user grip and control.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.