Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES 500 ml Blood Bag Canister, Baxter Fenwal 4R9955, Origen CS500, CyroMACS 200-074-402, Holds most 500 ml Blood Bags, 9 1/8L x 5 7/8D x 1/2H" (236 x 153 x 12mm) Constructed of lightweight aluminum.
Supplier:  CRYSTAL TECHNOLOGY AND INDUSTRIES C500
Encompass_Preferred
Canisters are designed with locking handle and thumb for easy handling. Many more sizes available to accommodate Blood Bags, Bone Marrow, Stem Cell, Cord Blood and Plasma Bag Storage. Custom Rack Design Services available.
Catalog No. NC2208114