Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
canine NKp46 (CD335) Mouse anti-Canine, Unconjugated, Clone: 48A, MilliporeSigma™
Mouse Monoclonal Antibody
$177.00 - $444.00
Specifications
Antigen | canine NKp46 (CD335) |
---|---|
Clone | 48A |
Applications | ELISA, Flow Cytometry, Immunocytochemistry |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF21092MI
|
MilliporeSigma
MABF210925UG |
25 μg |
Each for $177.00
|
|
|||||
MABF21091MI
|
MilliporeSigma
MABF2109100UG |
100 μg |
Each for $444.00
|
|
|||||
Description
canine NKp46 (CD335) Monoclonal antibody specifically detects canine NKp46 (CD335) in Canine samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry.Specifications
canine NKp46 (CD335) | |
ELISA, Flow Cytometry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Canine | |
Natural cytotoxicity triggering receptor 1;NK cell-activating receptor;Natural killer cell p46-related protein;NK-p46;Canine NKp46;CD335 | |
L-cells expressing canine NKp46 fusion protein | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
36.13 kDa calculated. |
48A | |
Monoclonal | |
Purified | |
Inflammation and Immunology | |
NP_001271377 | |
NCR1 | |
IgG2a κ | |
Protein G Purified | |
Clone 48A is a mouse monoclonal antibody that specifically detects canine NKp46. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title