Learn More
Enzo Life Sciences Canine coronavirus antigen, (native) (1mg)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Enzo Life Sciences ENZPRT3041000
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Formulation: Liquid. In 0.05M glycine, pH 9 with <0.1% CHAPS and <0.01% SDS. Source: Isolated from A-72 cells/Virus strain 1-71. Long Term Storage: -20°C. Applications: ELISA .