Learn More
Cancer Diagnostics, Inc.™ Vortex™ Biopsy Micro Cassettes
Used for routine processing and embedding of tissues and cells during histological studies
$209.90 - $318.00
Catalog Number | Mfr. No. | Color | Packaging Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Packaging Type | Price | Quantity & Availability | ||||
22-500-871
![]() |
Cancer Diagnostics, Inc.™
VM1002 |
Blue | Loose |
Case of 1000 for $305.53
|
|
22-500-870
![]() |
Cancer Diagnostics, Inc.™
VM1000 |
White | Loose |
Case of 1000 for $305.53
|
|
22-500-873
![]() |
Cancer Diagnostics, Inc.™
VM1004 |
Pink | Loose |
Case of 1000 for $305.53
|
|
22-500-872
![]() |
Cancer Diagnostics, Inc.™
VM1003 |
Green | Loose |
Case of 1000 for $305.53
|
|
22-500-874
![]() |
Cancer Diagnostics, Inc.™
VM1005 |
Yellow | Loose |
Case of 1000 for $305.53
|
|
22-500-875
![]() |
Cancer Diagnostics, Inc.™
VMT1000 |
White | Taped |
Case of 1000 for $318.00
|
|
22-500-876
![]() |
Cancer Diagnostics, Inc.™
VMT1002 |
Blue | Taped |
Case of 1000 for $314.85
|
|
22-500-877
![]() |
Cancer Diagnostics, Inc.™
VMT1003 |
Green | Taped |
Case of 1000 for $295.00
|
|
22-500-878
![]() |
Cancer Diagnostics, Inc.™
VMT1004 |
Pink | Taped |
Case of 1000 for $209.90
|
|
22-500-879
![]() |
Cancer Diagnostics, Inc.™
VMT1005 |
Yellow | Taped |
Case of 1000 for $295.00
|
|
22-500-880
![]() |
Cancer Diagnostics, Inc.™
VMSL1000 |
White | Tubed | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
22-500-884
![]() |
Cancer Diagnostics, Inc.™
VMSL1005 |
Yellow | Tubed | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
22-500-881
![]() |
Cancer Diagnostics, Inc.™
VMSL1002 |
Blue | Tubed | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
22-500-882
![]() |
Cancer Diagnostics, Inc.™
VMSL1003 |
Green | Tubed | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
22-500-883
![]() |
Cancer Diagnostics, Inc.™
VMSL1004 |
Pink | Tubed | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Rounded micro hole interior basket without corners
- Filtration enhanced by side vents
- Two-piece lid design
- Processed biopsies are easy to find
- Various colors available
- Tubed, loose and taped options for packaging
Specifications
Biopsy Micro Cassette |