Learn More
CAMPYLOBACTER QUIK CHEK™
Campylobacter infection is a leading cause of bacterial diarrhea. The specialized growth conditions for culturing this pathogen can be tedious and time-consuming. The CAMPYLOBACTER QUIK CHEK™ test offers excellent accuracy and rapid turnaround time without the downsides of culture.
Developed and Manufactured by TECHLAB, Inc.Supplier: Techlab Inc T31025
Description
The CAMPYLOBACTER QUIK CHEK™ test offers accurate diagnostic results in 30 minutes – culture can take up to 72 hours and is prone to false negative results due to stringent transport and growth conditions.
- Flexible specimen types include fresh, frozen, and Cary Blair and C&S transport media
- The CAMPYLOBACTER QUIK CHEK™ test detects C. jejuni, C. coli, C. upsaliensis, and C. lari
- Samples may be performed in batches or individually for on-demand testing
Specifications
C. jejuni, C. coli, C. upsaliensis, C. lari | |
Campylobacter QUIK CHEK™ | |
25 Membrane device pouches, diluent, wash buffer, conjugate, substrate, positive control, disposable plastic transfer pipettes | |
25 Tests/Kit |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.