Learn More
Roche Diagnostics Multi - Calibrator B Bio, 1 each
Supplier: Roche Diagnostics 06682553001
Set B consisting of a multi-calibrator and multi-controls for the Cedex Bio and Cedex Bio HT Systems serve for calibration and QC of the tests for Glutamine, and Ammonium
This item is not returnable.
View return policy