Learn More
Cole-Parmer Oakton Conductivity and TDS Standard, 84 µS; 500 mL
Supplier: Cole-Parmer 0065316
All Oakton Conductivity and TDS Standards are traceable to NIST standard reference materials. The solutions help you calibrate conductivity instruments to ensure accuracy with standard values given at 25C. Use when readings are expected to be around 84 S/cm.