Learn More
Bioworld Calf Serum, sterile filtered, 0.2 micron absolute, 500 mL
Supplier:  Bioworld 306111802
Encompass_Preferred
Sterility tested.Mycoplasma tested upon request.Endotoxin level: < 1.0 ng/ml.Sterile filtered to 0.2 µm.Storage Conditions: -20°C + 5°C.Can be heat inactivated if desired.Usage:bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical
Catalog No. 50-254-252