Learn More
Bioworld Calf Serum, sterile filtered, 0.2 micron absolute, 100 mL
Supplier: Bioworld 306111801
Sterility tested.Mycoplasma tested upon request.Endotoxin level: < 1.0 ng/ml.Sterile filtered to 0.2 µm.Storage Conditions: -20°C + 5°C.Can be heat inactivated if desired.Usage:bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.