Learn More
Colorado Serum Co Calf Alsevers 100mL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Colorado Serum Co 32033
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Recommend blood stored at 2-7 degrees C. Fragility increases after 21 days.