Learn More
Novex™ Calyculin A
Calculin A
Supplier: Novex™ PHZ1044
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Description
Calyculin A is a structurally unique marine toxin with potent and specific protein phosphatase inhibiting activity. This compound inhibits PP2A with similar potency to okadaic acid, but inhibits PP1 with a 10- to 100-fold greater potency.Format/Formulation: Lyophilized
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Store in freezer (-5 to -30°C). | |
5519 | |
TLC | |
PP2A and PP1 Phosphatases | |
Phosphatase Inhibitor |
1009.2 Da | |
Dry Ice | |
Non-sterile | |
Chemical | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.