Learn More
Hach Company Calcium Separation Set (for TNT852)
Supplier: Hach Company TNT892
The Calcium Separation Set is required when conducting TNTplus tests for Cadmium (TNT852) in sample matrices that contain calcium or magnesium in excess of 50 mg/L. Use of this set is in addition to digestion (using TNT890) when measuring total Cadmium.