Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium Citrate, Tetrahydrate, Powder, USP, 97.5-100.5%, Spectrum™ Chemical

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
125 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Calcium Citrate, Tetrahydrate, Powder, USP is used to treat conditions caused by low calcium levels such as bone loss (osteoporosis) and weak bones (osteomalacia/rickets). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 100% |
Loss on Drying | 10.0-13.3% |
Packaging | Fiber Drum |
Quantity | 125 lb. |
Percent Purity | 97.5-100.5% |
Grade | USP |