Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Calcium Carbide
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade
Description
Generate acetylene gas
This product is for educational use - no C of A is available.
Specifications
Specifications
Color | Black to Gray |
Quantity | 500 g |
Assay Percent Range | >90% |
Linear Formula | CaC2 |
Formula Weight | 64.08 |
Grade | Laboratory |
Packaging | Lab Grade |
Density | 2.22g/mL |
Chemical Name or Material | Calcium Carbide |
Safety and Handling
ShelfLife | 1 year |
DOTInformation | DOT Class 4.3, II, : Dangerous When Wet |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction