Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Calcitonin (8-32), salmon

Catalog No. AAJ64415LB0
Click to view available options
5 mg

Chemical Identifiers

CAS 155069-90-2
Molecular Formula C119H198N36O37
Molecular Weight (g/mol) 2725.11
MDL Number MFCD00671418
InChI Key CUQBOYYGZCRRKW-LPJWDPCBSA-N
Synonym Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH2;
IUPAC Name (4S)-4-{[(1S)-1-{[(1S)-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-{[(1S,2R)-1-{[(2S)-1-[(2S)-2-{[(1S)-4-carbamimidamido-1-{[(1S,2R)-1-{[(1S)-2-carbamoyl-1-{[(1S,2R)-1-[({[(1S)-1-[({[(2S,3R)-1-[(2S)-2-carbamoylpyrrolidin-1-yl]-3-hydroxy-1-oxobutan-2-yl]carbamoyl}methyl)carbamoyl]-2-hydroxyethyl]carbamoyl}methyl)carbamoyl]-2-hydroxypropyl]carbamoyl}ethyl]carbamoyl}-2-hydroxypropyl]carbamoyl}butyl]carbamoyl}pyrrolidin-1-yl]-3-(4-hydroxyphenyl)-1-oxopropan-2-yl]carbamoyl}-2-hydroxypropyl]carbamoyl}-3-carbamoylpropyl]carbamoyl}-3-methylbutyl]carbamoyl}pentyl]carbamoyl}-2-(1H-imidazol-4-yl)ethyl]carbamoyl}-3-methylbutyl]carbamoyl}-4-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-6-amino-2-{2-[(2S)-2-[(2S)-2-amino-3-methylbutanamido]-4-methylpentanamido]acetamido}hexanamido]-4-methylpentanamido]-3-hydroxypropanamido]-4-carbamoylbutanamido]butanoic acid
SMILES CC(C)C[C@H](NC(=O)[C@@H](N)C(C)C)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC1=CNC=N1)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC1=CC=C(O)C=C1)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H]([C@@H](C)O)C(=O)NCC(=O)N[C@@H](CO)C(=O)NCC(=O)N[C@@H]([C@@H](C)O)C(=O)N1CCC[C@H]1C(N)=O

Specifications

Physical Form Solid
Quantity 5 mg
Formula Weight 2725.06
ShelfLife 3 Years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.