Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ TMB Solution
Validated for use in ELISA for detection of TMB Solution. MilliporeSigma™; Calbiochem™ TMB Solution is soluble, non-toxic, non-flammable horseradish peroxidase substrate.
$157.00 - $566.00
Specifications
Form | Liquid |
---|---|
For Use With (Application) | ELISA |
pH | 3.55 ± 0.20 |
Product Type | TMB Solution |
Storage Requirements | Store at 2° to 8°C. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
CL07100ML
|
MilliporeSigma™
CL07100ML |
100mL |
Each for $157.00
|
|
|||||
CL071000ML
|
MilliporeSigma™
CL071000ML |
1000mL |
Each for $566.00
|
|
|||||
Description
- Form: Liquid
- Formulation: In a mildly acidic buffer with proprietary stabilizers, pH 3.55 ± 0.20
Specifications
Liquid | |
3.55 ± 0.20 | |
Store at 2° to 8°C. |
ELISA | |
TMB Solution |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title