Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ OmniPur™ IPTG
Inducer for β-galactosidase, an enzyme that promotes lactose utilization.
$121.70 - $1950.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Description | IPTG, Crystalline Powder |
Purity | Molecular Biology Grade |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
58-001-GM
|
MilliporeSigma™
58001GM |
1 g |
Each for $121.70
|
|
|||||
58-105-GM
|
MilliporeSigma™
58105GM |
5 g |
Each for $408.00
|
|
|||||
58-201-00GM
|
MilliporeSigma™
5820100GM |
100 g |
Each for $1,950.00
|
|
|||||
58-152-5GM
|
MilliporeSigma™
581525GM |
25 g | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
EMD Millipore Calbiochem™ OmniPur™ IPTG is used in conjunction with X-Gal.
- Molecular Biology grade
- Stock solutions:
- X-Gal: 20mg/mL in DMF
- IPTG: 20mg/mL in water
- Use 4:1 (X-Gal:IPTG)
Specifications
2°C to 8°C | |
IPTG, Crystalline Powder | |
Molecular Biology Grade |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title