Learn More
MilliporeSigma™ Calbiochem™ Collagen, Type I, Human Placenta
Used as gel or thin coating for attachment of cells
Supplier: MilliporeSigma™ 234149250UG
Description
- Highly purified, native, type I collagen from human placenta
Specifications
0.2 to 2 μg/cm2 of growth surface | |
250 μg | |
Collagen, Human Placenta Type I | |
Liquid |
In 10mM HCl | |
Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV. | |
-20°C | |
≥90% by SDS-PAGE |