Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Bovine γ-Globulin, Serum
Native γ-globulin from bovine serum.
$98.00 - $1148.00
Specifications
Content And Storage | Ok to freeze. |
---|---|
Form | Lyophilized |
Purity or Quality Grade | ≥97% by electrophoresis |
Species | Bovine serum |
Product Type | Adult Bovine Serum |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
34-587-65GM
|
MilliporeSigma™
3458765GM |
5 g |
Each for $98.00
|
|
|||||
34-587-625GM
|
MilliporeSigma™
34587625GM |
25 g |
Each for $397.00
|
|
|||||
34-587-6100GM
|
MilliporeSigma™
345876100GM |
100 g |
Each for $1,148.00
|
|
|||||
Description
- Form: Lyophilized
- Purity: ≥97% by electrophoresis
Specifications
Ok to freeze. | |
≥97% by electrophoresis | |
Adult Bovine Serum |
Lyophilized | |
Bovine serum |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title