Learn More
FIREBIRD OPTICS Calcium Fluoride (CaF2) Circular Optical Window- 25x4mm
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: FIREBIRD OPTICS CAF225X4
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Calcium Fluoride (CaF2) Circular Optical Window- 25x4mm• Wavelength range: 130nm-9.5µm •Surface quality: 60/40 • Clear aperture: 90% •Diameter tolerance: ±0.1mm •Thickness tolerance: ±0.2mm