Learn More
Electron Microscopy Sciences Cacodylic Acid-Sodium Salt, 124-65-2, MFCD00149079, 500 g
Supplier: Electron Microscopy Sciences 12310
Formula: C2H6AsNaO2.3H2O, MW: 214.02, 99.52%, Small white crystals, Names: Sodium dimethyl arsenate, TrihydrateSodium Cacodylate.