Learn More
Teledyne ISCO, Inc RediSep Rf Gold C18Aq, 50 g
Supplier: Teledyne ISCO, Inc 692203561
RediSep Rf Gold C18Aq is specifically designed for highly aqueous conditions. Monofunctionalized C18 bonding is interspersed with hydrophilic ligands to prevent phase collapse in high aqueous conditions. RediSep Rf Gold C18Aq should be used in separations requiring 0–50% organic. These columns offer increased retention through increased polar interactions and the ability to use weaker solvent system. This is usefulfor highly polar, water soluble compounds such as dyes, glycopeptides, and nucleotides.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.